Anti-OPN (secreted phosphoprotein 1) Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX37582
89282-420EA 671.04 USD
89282-420
Anti-OPN (secreted phosphoprotein 1) Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Osteopontin

P-ELISA:1:500-1000, WB:1:100-500, IHC-P:1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: OPN (secreted phosphoprotein 1)
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Bovine, Goat, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR