Anti-Reelin (reelin) Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX37552
89328-502EA 591.06 USD
89328-502
Anti-Reelin (reelin) Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Reelin

Recommended Starting Dilutions: For ICC/IF: Use at a dilution of 1:100-1:800. For IHC-P: Use at a dilution of 1:100-1:800. Optimal dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: Reelin (reelin)
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR