Anti-STAT5 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX37534
89277-228EA 606.05 USD
89277-228
Anti-STAT5 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to STAT5

P-ELISA:1:500-1000, WB:1:100-500, IHC-P:1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: STAT5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR