Anti-TGFBR2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX37527
89282-378EA 740.64 USD
89282-378
Anti-TGFBR2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to TGF beta Receptor 2

P-ELISA:1:500-1000, WB:1:100-500, IHC-P:1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: TGFBR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR