Anti-Dopey 2 (dopey Family member 2) Rabbit Polyclonal Antibody

GTX37489
89281-844EA 265.14 USD
89281-844
Anti-Dopey 2 (dopey Family member 2) Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to Dopey 2

WB:1:250-500

Type: Primary
Antigen: Dopey 2 (dopey Family member 2)
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR