Anti-REST Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX37363
89294-768EA 584.52 USD
89294-768
Anti-REST Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to NRSF

P-ELISA:1:500-1000, WB:1:100-500, IHC-P:1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: REST
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR