Anti-IL12 p40 (interleukin 12b) Rat Monoclonal Antibody

GTX37247
89282-318EA 426.21 USD
89282-318
Anti-IL12 p40 (interleukin 12b) Rat Monoclonal Antibody
Rat Monoclonal antibody to IL12 p40

Type: Primary
Antigen: IL12 p40 (interleukin 12b)
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR