Anti-AACT Mouse Monoclonal Antibody [clone: 81]

Supplier: GeneTex
GTX36929
89281-214EA 604.58 USD
89281-214
Anti-AACT Mouse Monoclonal Antibody [clone: 81]
Antibodies
Type: Primary
Antigen: AACT
Clonality: Monoclonal
Clone: 81
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR