Anti-O157 Mouse Monoclonal Antibody [clone: 1023]

Supplier: GeneTex
GTX36926
89332-142EA 645.27 USD
89332-142
Anti-O157 Mouse Monoclonal Antibody [clone: 1023]
Antibodies
Type: Primary
Antigen: O157
Clonality: Monoclonal
Clone: 1023
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR