Anti-O157 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 1031]

GTX36922
89332-134EA 376.83 USD
89332-134
Anti-O157 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 1031]
Type: Primary
Antigen: O157
Clonality: Monoclonal
Clone: 1031
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR