Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Parainfluenza virus type 1 Mouse Monoclonal Antibody [clone: 1226]

Supplier: Genetex
GTX36910
89332-120
Anti-Parainfluenza virus type 1 Mouse Monoclonal Antibody [clone: 1226]
Type: Primary
Antigen: Parainfluenza virus type 1
Clonality: Monoclonal
Clone: 1226
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus

Order Now


 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty