Anti-Parainfluenza virus type 3 Mouse Monoclonal Antibody [clone: 1254]

Supplier: GeneTex
GTX36906
89332-112EA 652.21 USD
89332-112
Anti-Parainfluenza virus type 3 Mouse Monoclonal Antibody [clone: 1254]
Antibodies
Type: Primary
Antigen: Parainfluenza virus type 3
Clonality: Monoclonal
Clone: 1254
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR