Anti-Influenza A virus NP Mouse Monoclonal Antibody [clone: 1371]

Supplier: GeneTex
GTX36899
89332-098EA 652.21 USD
89332-098
Anti-Influenza A virus NP Mouse Monoclonal Antibody [clone: 1371]
Antibodies
Type: Primary
Antigen: Influenza A virus NP
Clonality: Monoclonal
Clone: 1371
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR