Anti-Influenza A virus NP Mouse Monoclonal Antibody [clone: 1381]

Supplier: GeneTex
GTX36898
89332-096EA 652.21 USD
89332-096
Anti-Influenza A virus NP Mouse Monoclonal Antibody [clone: 1381]
Antibodies
Type: Primary
Antigen: Influenza A virus NP
Clonality: Monoclonal
Clone: 1381
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR