Anti-HADV Mouse Monoclonal Antibody [clone: 1421]

Supplier: GeneTex
GTX36896
89332-092EA 652.21 USD
89332-092
Anti-HADV Mouse Monoclonal Antibody [clone: 1421]
Antibodies
Type: Primary
Antigen: Adenovirus hexon
Clonality: Monoclonal
Clone: 1421
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR