Anti-Chlamydia trachomatis MOMP Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 1661]

GTX36879
89332-070EA 386.41 USD
89332-070
Anti-Chlamydia trachomatis MOMP Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 1661]
Type: Primary
Antigen: Chlamydia trachomatis MOMP
Clonality: Monoclonal
Clone: 1661
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR