Anti-Chlamydia trachomatis MOMP Mouse Monoclonal Antibody [clone: 1671]

Supplier: GeneTex
GTX36878
89332-068EA 395.3 USD
89332-068
Anti-Chlamydia trachomatis MOMP Mouse Monoclonal Antibody [clone: 1671]
Type: Primary
Antigen: Chlamydia trachomatis MOMP
Clonality: Monoclonal
Clone: 1671
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR