Anti-Chlamydia trachomatis LPS Mouse Monoclonal Antibody [clone: 1685]

Supplier: GeneTex
GTX36870
89332-060EA 652.21 USD
89332-060
Anti-Chlamydia trachomatis LPS Mouse Monoclonal Antibody [clone: 1685]
Antibodies
Type: Primary
Antigen: Chlamydia trachomatis LPS
Clonality: Monoclonal
Clone: 1685
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR