Anti-HBcAg Mouse Monoclonal Antibody [clone: 1823]

Supplier: GeneTex
GTX36863
89332-048EA 652.21 USD
89332-048
Anti-HBcAg Mouse Monoclonal Antibody [clone: 1823]
Antibodies
Type: Primary
Antigen: Hepatitis B Virus core antigen
Clonality: Monoclonal
Clone: 1823
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR