Anti-Hepatitis B virus santigen capture Mouse Monoclonal Antibody [clone: 1835]

Supplier: GeneTex
GTX36860
89332-042EA 652.21 USD
89332-042
Anti-Hepatitis B virus santigen capture Mouse Monoclonal Antibody [clone: 1835]
Antibodies
Type: Primary
Antigen: Hepatitis B virus santigen capture
Clonality: Monoclonal
Clone: 1835
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR