Anti-Hepatitis A virus Mouse Monoclonal Antibody [clone: 1885]

Supplier: GeneTex
GTX36854
89332-030EA 652.21 USD
89332-030
Anti-Hepatitis A virus Mouse Monoclonal Antibody [clone: 1885]
Antibodies
Type: Primary
Antigen: Hepatitis A virus
Clonality: Monoclonal
Clone: 1885
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR