Anti-HBeAg Mouse Monoclonal Antibody [clone: 1893]

Supplier: GeneTex
GTX36851
89332-024EA 652.21 USD
89332-024
Anti-HBeAg Mouse Monoclonal Antibody [clone: 1893]
Antibodies
Type: Primary
Antigen: Hepatitis B virus e antigen
Clonality: Monoclonal
Clone: 1893
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR