Anti-HIV1 p24 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 1941]

Supplier: GeneTex
GTX36848
89332-018EA 649.23 USD
89332-018
Anti-HIV1 p24 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 1941]
Antibodies
FITC-conjugated Mouse monoclonal antibody [1941] to HIV-1 p24

Recommended Starting Dilutions:For ICC/IF: Use at a starting dilution of 1:5 - 1:20. Acetone fixation is recommended.Not yet tested in other applications. Optimal dilutions should be determined experimentally by the researcher.<p>Best ELISA pairs: (capture/probe): 1941/1949, 1946/1949

Type: Primary
Antigen: HIV1 p24
Clonality: Monoclonal
Clone: 1941
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR