Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HIV1 p24 Mouse Monoclonal Antibody [clone: 1947]

Supplier: Genetex
GTX36847
Mouse monoclonal antibody [1947] to HIV-1 p24

Recommended Starting Dilutions:For ICC/IF: Use at 1:10-1:50For WB: Use at 1:10-1:50For IHC: Use at 1:10-1:50For ELISA: Use at 1:20-1:200Not yet tested in other applications. Optimal dilutions should be determined experimentally by the researcher.<p>Best ELISA pairs for this target: (capture/probe): 1941 / 1949, 1946 / 1949

Type: Primary
Antigen: HIV1 p24
Clonality: Monoclonal
Clone: 1947
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
500 μl GTX36847 89332-016 Each (500µl) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol HIV1 p24
Reactivity Virus
Conjugation Unconjugated
Clonality Monoclonal
Antibody Type Primary
Clone 1947
Host Mouse
Isotype IgG1
ELISA Yes
ImmunoFluorescence Yes
Concentration 100µg/ml
Purification Purified by protein A chromatography or sequential differential precipitations
Storage Buffer 0.01 M phosphate buffered saline, pH 7.2 and contains 0.1% sodium azide
Storage Temperature Store at -20°C until ready for use. Avoid repeated freeze-thawing by storing multiple aliquots at -20°C.
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty