Anti-HIV1 p24 Mouse Monoclonal Antibody [clone: 1948]

Supplier: GeneTex
GTX36846
89332-014EA 638.43 USD
89332-014
Anti-HIV1 p24 Mouse Monoclonal Antibody [clone: 1948]
Antibodies
Mouse monoclonal antibody [1948] to HIV-1 p24

Recommended Starting Dilutions:For ICC/IF: Use at 1:10-1:50For WB: Use at 1:10-1:50For IHC: Use at 1:10-1:50For ELISA: Use at 1:20-1:200Not yet tested in other applications. Optimal dilutions should be determined experimentally by the researcher.<p>Best ELISA pairs for this target: (capture/probe): 1941 / 1949, 1946 / 1949

Type: Primary
Antigen: HIV1 p24
Clonality: Monoclonal
Clone: 1948
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR