Anti-HIV1 GP120 Mouse Monoclonal Antibody [clone: 1994]

GTX36844
89332-010EA 376.81 USD
89332-010
Anti-HIV1 GP120 Mouse Monoclonal Antibody [clone: 1994]
Mouse monoclonal antibody [1994] to HIV-1 gp120

Recommended Starting Dilutions:For ICC/IF: Use at 1:10-1:50For WB: Use at 1:10-1:50For IHC: Use at 1:10-1:50For ELISA: Use at 1:20-1:200Not yet tested in other applications. Optimal dilutions should be determined experimentally by the researcher.<p>Best ELISA pairs for this target: (capture/probe): 1941 / 1949, 1946 / 1949

Type: Primary
Antigen: HIV1 GP120
Clonality: Monoclonal
Clone: 1994
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR