Anti-Treponema pallidum p47 Mouse Monoclonal Antibody [clone: 2131]

Supplier: GeneTex
GTX36836
89332-006EA 395.25 USD
89332-006
Anti-Treponema pallidum p47 Mouse Monoclonal Antibody [clone: 2131]
Type: Primary
Antigen: Treponema pallidum p47
Clonality: Monoclonal
Clone: 2131
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR