Anti-Canine Heartworm Mouse Monoclonal Antibody [clone: 3233]

Supplier: GeneTex
GTX36831
89331-996EA 652.21 USD
89331-996
Anti-Canine Heartworm Mouse Monoclonal Antibody [clone: 3233]
Antibodies
Type: Primary
Antigen: Canine Heartworm
Clonality: Monoclonal
Clone: 3233
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR