Anti-Francisella tularensis LPS Mouse Monoclonal Antibody [clone: 3515]

Supplier: GeneTex
GTX36828
89331-990EA 482.84 USD
89331-990
Anti-Francisella tularensis LPS Mouse Monoclonal Antibody [clone: 3515]
Antibodies
Type: Primary
Antigen: Francisella tularensis LPS
Clonality: Monoclonal
Clone: 3515
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR