Anti-Brucella abortus LPS Mouse Monoclonal Antibody [clone: 3535]

Supplier: GeneTex
GTX36825
89331-984EA 668.53 USD
89331-984
Anti-Brucella abortus LPS Mouse Monoclonal Antibody [clone: 3535]
Antibodies
Type: Primary
Antigen: Brucella abortus LPS
Clonality: Monoclonal
Clone: 3535
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR