Anti-Plasmodium falciparum MSP1 Mouse Monoclonal Antibody [clone: 3566]

Supplier: GeneTex
GTX36819
89331-972EA 482.84 USD
89331-972
Anti-Plasmodium falciparum MSP1 Mouse Monoclonal Antibody [clone: 3566]
Antibodies
Type: Primary
Antigen: Plasmodium falciparum MSP1
Clonality: Monoclonal
Clone: 3566
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Protozoa
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR