Anti-Enterovirus 70 Mouse Monoclonal Antibody [clone: 3651]

Supplier: GeneTex
GTX36809
89331-958EA 652.21 USD
89331-958
Anti-Enterovirus 70 Mouse Monoclonal Antibody [clone: 3651]
Antibodies
Type: Primary
Antigen: Enterovirus 70
Clonality: Monoclonal
Clone: 3651
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR