Anti-West Nile Virus Mouse Monoclonal Antibody [clone: 3671]

Supplier: GeneTex
GTX36805
89331-950EA 652.21 USD
89331-950
Anti-West Nile Virus Mouse Monoclonal Antibody [clone: 3671]
Antibodies
Type: Primary
Antigen: West Nile Virus
Clonality: Monoclonal
Clone: 3671
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR