Anti-SARS coronavirus nucleoprotein Mouse Monoclonal Antibody [clone: 3851]

Supplier: GeneTex
GTX36802
89331-944EA 634.97 USD
89331-944
Anti-SARS coronavirus nucleoprotein Mouse Monoclonal Antibody [clone: 3851]
Antibodies
Type: Primary
Antigen: SARS coronavirus nucleoprotein
Clonality: Monoclonal
Clone: 3851
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR