Anti-Clostridum difficile toxin A Mouse Monoclonal Antibody [clone: 5157]

Supplier: GeneTex
GTX36778
89331-904EA 652.21 USD
89331-904
Anti-Clostridum difficile toxin A Mouse Monoclonal Antibody [clone: 5157]
Antibodies
Type: Primary
Antigen:
Clonality: Monoclonal
Clone: 5157
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR