Anti-Mycobacterium tuberculosis Hsp65 Mouse Monoclonal Antibody [clone: 5177]

Supplier: GeneTex
GTX36770
89331-888EA 652.21 USD
89331-888
Anti-Mycobacterium tuberculosis Hsp65 Mouse Monoclonal Antibody [clone: 5177]
Antibodies
Type: Primary
Antigen: Mycobacterium tuberculosis Hsp65
Clonality: Monoclonal
Clone: 5177
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR