Anti-Legionella pneumophila Mouse Monoclonal Antibody [clone: 6027]

Supplier: GeneTex
GTX36761
89331-878EA 652.21 USD
89331-878
Anti-Legionella pneumophila Mouse Monoclonal Antibody [clone: 6027]
Antibodies
Type: Primary
Antigen: Legionella pneumophila
Clonality: Monoclonal
Clone: 6027
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR