Anti-Legionella pneumophila Mouse Monoclonal Antibody [clone: 6029]

Supplier: GeneTex
GTX36760
89331-876EA 652.21 USD
89331-876
Anti-Legionella pneumophila Mouse Monoclonal Antibody [clone: 6029]
Antibodies
Type: Primary
Antigen: Legionella pneumophila
Clonality: Monoclonal
Clone: 6029
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR