Anti-T. gondii Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 6201]

Supplier: GeneTex
GTX36750
89331-856EA 629.3 USD
89331-856
Anti-T. gondii Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 6201]
Antibodies
Type: Primary
Antigen: T. gondii
Clonality: Monoclonal
Clone: 6201
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype:
Reactivity: Protozoa
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR