Anti-Tg 38kDa Mouse Monoclonal Antibody [clone: 6205]

Supplier: GeneTex
GTX36748
89331-852EA 652.21 USD
89331-852
Anti-Tg 38kDa Mouse Monoclonal Antibody [clone: 6205]
Antibodies
Type: Primary
Antigen: Tg 38kDa
Clonality: Monoclonal
Clone: 6205
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Protozoa
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR