Anti-T. gondii Mouse Monoclonal Antibody [clone: 6207]

Supplier: GeneTex
GTX36747
89331-850EA 652.21 USD
89331-850
Anti-T. gondii Mouse Monoclonal Antibody [clone: 6207]
Antibodies
Type: Primary
Antigen: T. gondii
Clonality: Monoclonal
Clone: 6207
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Protozoa
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR