Anti-Giardia lamblia 65 kDa Mouse Monoclonal Antibody [clone: 6261]

Supplier: GeneTex
GTX36744
89331-844EA 652.21 USD
89331-844
Anti-Giardia lamblia 65 kDa Mouse Monoclonal Antibody [clone: 6261]
Antibodies
Type: Primary
Antigen: Giardia lamblia 65 kDa
Clonality: Monoclonal
Clone: 6261
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Protozoa
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR