Anti-Salmonellae sp. common core Mouse Monoclonal Antibody [clone: 6394]

Supplier: GeneTex
GTX36734
89331-824EA 652.21 USD
89331-824
Anti-Salmonellae sp. common core Mouse Monoclonal Antibody [clone: 6394]
Antibodies
Type: Primary
Antigen: Salmonellae sp. common core
Clonality: Monoclonal
Clone: 6394
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR