Anti-Mycoplasma hominis p120 Mouse Monoclonal Antibody [clone: 6507]

Supplier: GeneTex
GTX36732
89331-820EA 652.21 USD
89331-820
Anti-Mycoplasma hominis p120 Mouse Monoclonal Antibody [clone: 6507]
Antibodies
Type: Primary
Antigen: Mycoplasma hominis p120
Clonality: Monoclonal
Clone: 6507
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR