Anti-Helicobacter pylori 58 kDa Mouse Monoclonal Antibody [clone: 6661]

Supplier: GeneTex
GTX36715
89331-788EA 652.21 USD
89331-788
Anti-Helicobacter pylori 58 kDa Mouse Monoclonal Antibody [clone: 6661]
Antibodies
Type: Primary
Antigen: Helicobacter pylori 58 kDa
Clonality: Monoclonal
Clone: 6661
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR