Anti-Trichomonas vaginalis Mouse Monoclonal Antibody [clone: 7151]

GTX36710
89331-780EA 366.15 USD
89331-780
Anti-Trichomonas vaginalis Mouse Monoclonal Antibody [clone: 7151]
Type: Primary
Antigen: Trichomonas vaginalis
Clonality: Monoclonal
Clone: 7151
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Protozoa
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR