Anti-Trichomonas vaginalis Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 7181]

Supplier: GeneTex
GTX36706
89331-772EA 635.92 USD
89331-772
Anti-Trichomonas vaginalis Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 7181]
Antibodies
Type: Primary
Antigen: Trichomonas vaginalis
Clonality: Monoclonal
Clone: 7181
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype:
Reactivity: Protozoa
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR