Anti-Bacillus anthracis spore antigen Mouse Monoclonal Antibody [clone: 7827]

Supplier: GeneTex
GTX36679
89331-744EA 528.06 USD
89331-744
Anti-Bacillus anthracis spore antigen Mouse Monoclonal Antibody [clone: 7827]
Antibodies
Type: Primary
Antigen: Bacillus anthracis spore antigen
Clonality: Monoclonal
Clone: 7827
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR