Anti-Staph. Enterotoxin toxin A Mouse Monoclonal Antibody [clone: 7935]

Supplier: GeneTex
GTX36675
89331-740EA 652.21 USD
89331-740
Anti-Staph. Enterotoxin toxin A Mouse Monoclonal Antibody [clone: 7935]
Antibodies
Type: Primary
Antigen: Staph. Enterotoxin toxin A
Clonality: Monoclonal
Clone: 7935
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR