Anti-Staph. Enterotoxin toxin B Mouse Monoclonal Antibody [clone: 7941]

Supplier: GeneTex
GTX36673
89331-736EA 652.21 USD
89331-736
Anti-Staph. Enterotoxin toxin B Mouse Monoclonal Antibody [clone: 7941]
Antibodies
Type: Primary
Antigen: Staph. Enterotoxin toxin B
Clonality: Monoclonal
Clone: 7941
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR